Learn More
Drucker™ Accessories for SampleSafe™ Storage Lock Box
Optional accessories for use with SampleSafe Storage Lock Box
Supplier: Drucker™ 03100120053
Specifications
Cold Pack | |
6 x 9 in. W x D, 0.8 in. thick. Aids in preserving sample integrity in warm environments. Fully tested to provide ideal storage for up to 10 hours. | |
Drucker SampleSafe LB1, LB10, LB2 and LB20 Lock Boxes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.