Learn More
Cole-Parmer Drierite 26800 Drying column, 200 L/hr, 90 psi
Supplier:  Cole-Parmer 0719300
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Polycarbonate drying column dries air and gas to a dew point of –100°F at up to 90 psi. Connect the column in-line between a dryer or desiccator and a compressed air line or gas cylinder; handles a flow of 200 liters per hour or 0.1 cfm. Measures 11-3/8"H x 2-5/8" dia. Column is made up of a plastic cap, 303 stainless steel desiccant supports and coil springs, felt filters, and O-ring gasket. Inlet and outlet ports accept 1/4" to 3/8" ID tubing. Absorbs up to 50 g of moisture. Drying column comes filled with 1-1/4 lb of 8 mesh indicating desiccant (anhydrous calcium sulfate). Order additional desiccant separately at left.
Catalog No. NC9760623