Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ DryRigger CoolRigger Oil and Water Resistant Gloves

Catalog No. p-7113041 Shop All Impacto Protective Products Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves repel oil and water, while also providing air flow to keep hands cool on hot summer days. TPR padding helps protect against hairline fractures, bruising, and pinched fingers.

  • Breathable backing material resists moisture while still allowing air flow to keep hands comfortable in hot conditions
  • Heavy duty TPR pad across knuckles, back of fingers, thumb & back of the hand protects against bruises & bumps
  • PVC nitrile layer offers an additional barrier of protection in oily conditions
  • Silicone dots on the palm and fingertips enhance grip capabilities
  • Ultra suede fabric in the palm repels oil & water
  • Durable patches in the webbing area assure long wear

Specifications

Product Type Anti-Impact Gloves
Back Material TPR Padding
Color Blue, Yellow
Cuff Material Neoprene
For Use With (Application) Mining, Oil/Gas Extraction, and Well Drilling
Liner Material PVC Nitrile
Material Suede
Palm Type Coated
Resistant To Oil, Water
Series DryRigger CoolRigger
Size Small
Glove Cuff Color Black
EN388 Standard 3231
Coating Material Silicone
Hand Specific
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.