Learn More
AdipoGen DSP-1
SDP
Supplier:  AdipoGen CDXD0391M010
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 1609395-29-0. Formula C28H16FNO8. MW 494.44. Synthetic Fluorescent probe for detection of H2S2 and hydrogen polysulfides in aqueous buffers.
Catalog No. 50-256-1093