Learn More
AdipoGen DSP-2
SDP
Supplier:  AdipoGen CDXD0392M005
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 1609395-31-4. Formula C27H14FNO8. MW 499.4. Synthetic Control for H2S2 and hydrogen polysulfides probes.
Catalog No. 50-256-1091
missing translation for 'QuantityLeft'