Learn More
AdipoGen DSP-3
SDP
Supplier:  AdipoGen CDXD0393M005
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 1609395-32-5. Formula C34H16F2N2O11. MW 666.49. Synthetic Cell-permeable fluorescent probe for sensitive detection limit ~71 nM and selective detection of H2S2 and hydrogen polysulfides in aqueous buffers as well as in cells/biological systems. Shows almost no cytotoxicity and can be used in detecting H2Sn not H2S in cells.
Catalog No. 50-256-1088
missing translation for 'QuantityLeft'