Learn More
Euthanex Pole-Mounted Dual Oxygen Tank Manifold with Regulator and Pressure Gauge. Used in conjunction with the EZ-CRVM Dual Cylinder Oxygen Tanks Holder.
Supplier:  Euthanex EZDOM
Encompass_Preferred
Pole-Mounted Dual Oxygen Tank Manifold with Regulator and Pressure Gauge. Used in conjunction with the EZ-CRVM Dual Cylinder Oxygen Tanks Holder.
Catalog No. 50-154-0188
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.