Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ DualFusion Vials

Catalog No. p-7076783 Shop All DWK Life Sciences Products
Click to view available options
Type:
Volume (Metric):
No. per Case:

For storage and transport of samples. Wheaton™ DualFusion Vials maintains sample integrity and protect sensitive reagents.

  • Combines best properties of both plastic and glass into one vial
  • “FUSION” material lining provides protection against leachates, delamination, and gas permeation
  • Cyclic Olefin Polymer (COP) shell offers the comfort of a break resistant vial
  • Manufactured in a Class 7 cleanroom so users receive a “ready-to-sterilize” vial
  • Equipped with a 2D bar code which provides complete manufacturing traceability
  • Can be sterilized via Gamma Radiation, ETO and E-beam

Compliance

Meet USP<788> for low particulate levels and USP<85> for low endotoxin level for injectables

Specifications

Diameter (Metric) Inner

22 mm

Color

Clear

Volume (Metric)

5 mL

Liner Material

FUSION

No. per Case

48

Temperature Range (Metric)

-196°C to +121°C

Closure Type

Crimp Top

Height (Metric)

40 mm

Certifications/Compliance

Meet USP<788> for low particulate levels and USP<85> for low endotoxin level for injecta

Type

DualFusion Vial 6 R

Closure Size

20 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.