Learn More
R&D Systems™ DuoSet ELISA Ancillary Reagent Kit 3
Shop All R&D Systems ProductsSpecifications
Product Type
Solid Phase Sandwich ELISA
For Use With (Application)
ELISA
Quantity
1 Kit (for 5 Plates)
Storage Requirements
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.