Learn More
Electron Microscopy Sciences Duplex Reference Specimen
Supplier:  Electron Microscopy Sciences 7952101
Encompass_Preferred
An alternative and very sensitive test is by means of an alloy with two major Copper/Zinc phases separated by an atomic number difference of 0.1. The light phase illustrated in the micrograph has a mean atomic number of 29.47 and the dark phase a mean atomic number of 29.37.
Catalog No. 50-248-18
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.