Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DY131, Tocris Bioscience™

Catalog No. p-4783929 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Agonist selective for estrogen-related receptors ERRβ and ERRγ

Novel selective agonist at estrogen-related receptors ERRβ and ERRγ. Displays minimal activity at ERRα, ERα and ERβ at concentrations up to 30 μM.

Chemical Identifiers

CAS 95167-41-2
Molecular Formula C18H21N3O2
Molecular Weight (g/mol) 311.385
InChI Key WLKOCYWYAWBGKY-CPNJWEJPSA-N
Synonym unii-5vwv92039e, n-4-diethylaminobenzylidenyl-n'-4-hydroxybenzoyl-hydrazine, d06jpq, n'-4-diethylamino phenyl methylidene-4-hydroxybenzohydrazide, n-e-4-diethylamino phenyl methylideneamino-4-hydroxybenzamide, benzoic acid, p-hydroxy-, p-diethylamino benzylidene hydrazide, n'-1e-4-diethylamino phenyl methylene-4-hydroxybenzohydrazide, benzoic acid, 4-hydroxy-, 4-diethylamino phenyl methylene hydrazide, benzoic acid, 4-hydroxy-, 2-4-diethylamino phenyl methylene hydrazide, dy hplc
PubChem CID 5497124
IUPAC Name N-[(E)-[4-(diethylamino)phenyl]methylideneamino]-4-hydroxybenzamide
SMILES CCN(CC)C1=CC=C(C=C1)C=NNC(=O)C2=CC=C(C=C2)O

Specifications

Quantity 10 mg
Formula Weight 311.38
Percent Purity >99%
Chemical Name or Material DY131
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.