Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dysprosium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific™

Catalog No. p-7051515
Click to view available options
Quantity:
10 g
50 g

For vacuum deposition

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 13569-80-7
MDL Number MFCD00016071

Specifications

Boiling Point 2200°C
Melting Point 1360°C
Color White
Physical Form Pieces
Quantity 10 g
Solubility Information Soluble in strong mineral acids. Insoluble in water.
Formula Weight 219.5
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 7.5 g/mL
Assay (REO)
Chemical Name or Material Dysprosium(III) fluoride, Anhydrous
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-992-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.