Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dysprosium(III) iodide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is used in high-intensity metal-halide lamps.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Specifications
Melting Point | 955°C |
Color | Green to Yellow |
Physical Form | Flakes |
Quantity | 1 g |
Solubility Information | Soluble in water. |
Formula Weight | 543.21 |
Percent Purity | 99.99% |
Packaging | Ampule |
Sensitivity | Hygroscopic |
Assay | (REO) |
Show More |
Safety and Handling
Hazard Category | H317-H334 |
Hazard Statement | GHS H Statement H334-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. |
Precautionary Statement | P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c |
EINECSNumber | 239-493-4 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction