Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein 98.0+%, TCI America™
SDP

Catalog No. p-7129918 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 93957-50-7
Molecular Formula C20H18FNO
Molecular Weight (g/mol) 307.368
MDL Number MFCD03840863
InChI Key DVWHSTKQJBIYCK-VMPITWQZSA-N
Synonym (E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]propenal
PubChem CID 9926593
IUPAC Name (E)-3-[3-(4-fluorophenyl)-1-propan-2-ylindol-2-yl]prop-2-enal
SMILES CC(C)N1C2=CC=CC=C2C(=C1C=CC=O)C3=CC=C(C=C3)F

Specifications

Melting Point 131°C
Color Yellow
Quantity 5 g
UN Number 3077
Formula Weight 307.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.