Learn More
Electron Microscopy Sciences E-Beam Lithography Mount
Supplier:  Electron Microscopy Sciences 7594415
Encompass_Preferred
This mount, made from aluminum features an M4 threaded hole and is 25 mm in diameter. The clips are made from copper. The mount comes complete with a Faraday Cup (2.5 mm x 100nm hole) and a gold/carbon resolution sample 3 mm in diameter is included. The gold particle size range is 5-150nm.
Catalog No. 50-192-8204
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.