Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis System, EX 2%, and E1-ClipTip™ Bluetooth™ Equalizer Pipette Package
SDP

Catalog No. A39485
Click to view available options
:
1 Package

This package combines everything you need to streamline your DNA sample analysis workflow and increase productivity.

This package combines everything you need to streamline your DNA sample analysis workflow and increase productivity.

The kit package includes:

  • E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
  • E1-ClipTip Bluetooth Electronic Adjustable Tip-Spacing Multichannel Equalizer Pipette (Cat. No. 4672040BT)
  • 2% E-Gel EX Agarose Gels, 10 gels (Cat. No. G401002)
  • E-Gel 50-bp DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488099)
  • Gel opening knife (Cat. No. EI9010)
  • ClipTip Filtered 384 Format Pipette Tips, 30 μL (Cat. No. 94420103)

E-Gel Power Snap Electrophoresis System

The all-in-one Invitrogen E-Gel Power Snap Electrophoresis System is designed for fast and convenient E-Gel agarose gel electrophoresis, from DNA sample loading to gel image. With dry precast E-Gel agarose gel technology, you can run DNA samples in as little as 10 minutes and observe sample separation in real time. It is the only benchtop system that seamlessly integrates DNA sample separation and gel imaging into one workflow.

Features of the E-Gel Power Snap Electrophoresis system include:

  • Streamline electrophoresis and image capture with an all-in-one system
  • Separate DNA in as little as 10 minutes with dry precast E-Gel agarose gels
  • View and capture DNA samples in real time
  • Designed for efficient and convenient benchtop use

E1-ClipTip Bluetooth Electronic Adjustable Tip-Spacing Multichannel Equalizer Pipette

Perform sample transfers between virtually any tube, rack, microplate, or horizontal gel box quickly and efficiently with the Thermo Scientific E1-ClipTip equalizer pipette. Adjustable tip spacing allows you to set the distance between tips by simply sliding the scale to expand or contract to accomodate the desired labware format. The My Pipette Creator app enables programming of the E1-ClipTip pipette from the comfort of your PC.

Specifications

Content And Storage

• E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
• E1-ClipTip Bluetooth Electronic Adjustable-Tip Spacing Multichannel Equalizer Pipette (Cat. No. 4672040BT)
• 10 E-Gel EX Agarose Gels, 2% (Cat. No. G401002)
• E-Gel 50-bp DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488099)
• Gel opening knife (Cat. No. EI9010)
• ClipTip Filtered 384 Format Pipette Tips, 30 μL (Cat. No. 94420103)

Storage: E-Gel Agarose Gels should be stored strictly at room temperature.

Product Line E-Gel
Type Horizontal Gel Electrophoresis System
Gel Percentage 2%
Gel Type E-Gel
Label or Dye Proprietary Stain
No. of Wells 11
Quantity 1 package
Shipping Condition Room Temperature
Well Design Single Comb

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.