Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ EasyPlus™ Polypropylene Tube Holder

Catalog No. 13940092 Shop All Mettler Toledo Products
Click to view available options
:

Accessory for EasyPlus™ Titrator

Securely holds the tube of the EasyPlus™ Easy Dose dosing unit and clips to the designated bottle, beaker, or flask being dosed into.

Specifications

Color

Clear

Description

Tube Holder

For Use With (Application)

Dosing

Includes

3 pieces

Material

Polypropylene

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.