Learn More
STEMCELL Technologies EasySep™ Human CD4+ T Cell Enrichment Kit, For processing: 1 x 10^9 cells
The EasySep™ Human CD4+ T Cell Enrichment Kit is designed to isolate CD4+ T cells from fresh or previously frozen peripheral mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-CD4+ T cells and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube. For even faster cell isolations, we recommend the new EasySep™ Human CD4+ T Cell Isolation Kit (17952) which isolates cells in just 8 minutes. Products are for Research Use Only, Not for Diagnostic or Therapeutic use, and are not to be administered to humans.