Learn More
Bioanalytical Systems Inc EC CGME CTRL DROP MERC ELECT
SDP
Supplier:  Bioanalytical Systems Inc EF1400
Encompass_Preferred
Electrode; Mercury Drop; BSI Controlled Growth; High overpotential for H ion reduction; High surface reproducibility; Sealed reservoir; Minimal use of metal parts; Three cell modes; Contains mercury ***SUPPLIER WILL ONLY SHIP TO PUERTO RICO! *** Do not place order if not located in Puerto Rico
Catalog No. 50-854-254
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.