Learn More
Bioanalytical Systems Inc EC PT WIRE AUXILLARY ELECT
SDP
Supplier:  Bioanalytical Systems Inc MW1032
Encompass_Preferred
Electrode, Platinum Wire Auxiliary; 0.5mm dia. x 7.5cm L; With gold-plated connector; Mounted in CTFE cylinder; For C-2 and C-3 cell stands and controlled growth mercury electrode *Does not ship to Puerto Rico
Catalog No. 50-854-892
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.