Learn More
Torrey Pines Scientific™ EchoTherm™ Benchtop Chilling/Heating Incubator Sample Blocks
Catalog No. p-151607
Click to view available options
Holds:
0.2mL tubes
0.5mL/1.5mL tubes
1.5mL tubes
Blocks are machined from solid aluminum to fit precisely the tube size and shape. Block bottoms are also machined to nest on the incubator plate to prevent movement. Feature an anodized black finish for better heat transfer.
Specifications
For Use With (Equipment) Echotherm benchtop chilling/heating incubator
Holds 0.2mL tubes
Material Aluminum
Type Block
For Use With (Application) For 0.2mL centrifuge tubes spaced for 8 and 12 tube strip
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.