Learn More
TARGETMOL CHEMICALS INC ECOPIPAM 5MG
Supplier:  TARGETMOL CHEMICALS INC T31602
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Ecopipam (UNII-0X748O646K) is a potent selective and orally active antagonist of dopamine D1/D5 receptor with Kis of 1.2 nM and 2.0 nM respectively. Ecopipam shows more than 40-flod selectivity over D2 D4 5-HT and (alpha)2a receptor (Ki=0.98 5.52 0.08 and 0.73 uM respectively). Ecopipam can be used for the research of schizophrenia and obesity. purity: 98%
Catalog No. 50-258-0362
missing translation for 'QuantityLeft'