Learn More
Edvotek™ HexaGel™ Electrophoresis Sixth-Tooth Comb FSE
SDPSCI_ED
Catalog No. S94649 Shop All EDVOTEK Products
Click to view available options
:
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
Six-tooth well-former comb
Well-former comb.
Polycarbonate comb casting six wells accommodates up to 38 to 40μL of sample
Specifications
For Use With (Equipment)
HexaGel™ Electrophoresis System
Type
Comb
Includes
6-teeth well-former comb and three E-Z Align™ gel casting tray
Teeth Count
6
Volume (Fill) Metric
38 to 40 μL