Learn More
ATCC Synthetic Eastern equine; encephalitis virus RNA
Supplier:  ATCC VR3239SD
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation, and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load. Preparation includes fragments from capsid, envelope (E1 and E2), and nonstructural genes NSP1, NSP3, and 3’UTR.
Catalog No. NC2205216
missing translation for 'QuantityLeft'