Learn More
TARGETMOL CHEMICALS INC EFONIDIPINE 25MG
Supplier:  TARGETMOL CHEMICALS INC T2323
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Efonidipine (NZ-105)(NZ-105) is a dihydropyridine calcium channel blocker blocking both T-type and L-type calcium channels. purity: 99%
Catalog No. 50-257-7161
missing translation for 'QuantityLeft'