Learn More
AG Scientific Inc EGTA 0.5M Solution pH 8.0,50ML
Supplier:  AG Scientific Inc E249150ML
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
EGTA 0.5M Solution is a useful chemical in a variety of buffers. EGTA is a chelating agent that can be used for the determination of calcium in the presence of magnesium since EGTA has greater specificity for calcium ions. EGTA is also useful in ELISA reactions as an inhibitor of alkaline phosphatase activity. EGTA has also been shown to activate Ca2+-stimulated ATPase in rat liver plasma membranes and induce apoptosis in PC12 cells. It also inhibits the zinc-dependent endopeptidases, MMPs (Matrix metalloproteinases).CAS Number: 67-42-5Molecular Weight: 380.35 DaChemical Formula: C14H24N2O10Solubility: Soluble in 1M NaOH (100 mM), Slightly soluble in water.Storage Temperature: Room TemperatureResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6999