Learn More
Gold Biotechnology Inc EGTA 100 g
SDP
Supplier:  Gold Biotechnology Inc E217100
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Ethyleneglycol bis(2-aminoethyl ether)-N,N,N',N' tetraacetic acid, (EGTA) is a calcium specific chelator. Because of its low affinity for Mg+, EGTA inhibits metalloproteases.
Catalog No. NC2943341
missing translation for 'QuantityLeft'