Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EHNA hydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective inhibitor of cGMP-stimulated phosphodiesterase (PDE2) (IC50 = 0.8 - 4 μM). Also a potent inhibitor of adenosine deaminase. Suppresses spontaneous differentiation of human ESCs in feeder-free conditions. Also prevents directed neuronal differentiation.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 313.83 |
Percent Purity | >98% |
Chemical Name or Material | EHNA hydrochloride |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction