Learn More
Hach Company Radiometer Analytical PHC2015-8 Combination Red-Rod pH Electrode for Alkaline Samples (alkaline glass, epoxy body, BNC)
Supplier:  Hach Company E16M318
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
The Radiometer PHC2015-8 Combination pH Electrode has a robust epoxy body, Red-Rod reference system, refillable electrolyte (sat. KCl) and porous pin junction. The PHC2015-8 has a glass membrane specific for alkaline samples. The PHC2015-8 is compatible with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).
Catalog No. 50-199-7580