Learn More
Cole-Parmer Oakton by Cole-Parmer self-cleaning pH electrode, Ryton housing, 1 k Ohms RTD
Supplier:  Cole-Parmer 2700324
Encompass_Preferred
There will be no further buildups with this self-cleaning electrode cable. Its flat surface prevents buildups, while fluid flow cleans surface for longer life. It can be used with most meters and controllers. The BNC connector and ATC element work with a variety of units. Its double junction is for difficult applications.
Catalog No. 50-633-488
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.