Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Electronic Rechargeable Crimpers and Decappers with OLED Display Screen

Hundreds of operations from one charge
Supplier: Restek™ 23237
Description
- OLED screen display to optimize crimping power for each cap type, monitor battery use and view lifetime statistics
- Large battery allows for faster charge cycle and two sleep mode options extend battery life
- Easy to use
- Comfortable grip
- All kits include a universal plug set for battery charger
- For use with 11 mm and 20 mm caps
- One year warranty
Specifications
Decapper | |
11 mm | |
OLED display screen | |
1 year |
Caps | |
6.4 V | |
Charger USB drive and universal plug set with U.S., European, UK and Australian plug clips |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction