Learn More
Agilent Technologies ELECTROTEN-BLUE E.COMP CELLS
Supplier:  Agilent Technologies 200159
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
ElectroTen-Blue Electroporation-Competent Cells 5 x 0.1mL and pUC18 control plasmid Highest electroporation efficiency (Hee) phenotype use for extrememly demanding cloning= 1 x 1010 transformants/microgram
Catalog No. 50-125-022
missing translation for 'QuantityLeft'