Learn More
ACROBiosystems ACROBiosystems ELISA Assay Kit for Anti-PD-1 h-mAb in Mouse Serum
Supplier:  ACROBiosystems EPMV196TESTS
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Kits;96TESTS;The enclosed ELISA assay kit for anti-PD-1 h-mAb in mouse serum is based on an enzyme immunoassay (ELISA) between recombinant PD-1 protein (ECD) and a biotinylated anti-PD-1 monoclonal antibody. The method employs the principle of competitive ELISA, allowing quantification of monoclonal anti-PD-1 antibodies in mouse serum.
Catalog No. 50-210-6633
missing translation for 'QuantityLeft'