Learn More
Biomatik Corporation Phospho-Synaptotagmin (Thr202) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50994
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Phospho-Synaptotagmin (Thr202) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Synaptotagmin (Thr202) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P21579; Q8N9I0. Gene ID: 6857; 127833. Target Synonyms: Synaptotagmin-1; Synaptotagmin I; SytI; p65; SYT1; SVP65; SYT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1206