Learn More
ELiTech Molecular Diagnostics MGB Alert™ Primers
Provide molecular labs with a more advanced alternative to develop real-time PCR-based assays
Supplier:  Nanogen, Inc 800350
 View more versions of this product
Catalog No. 22-801-067
This item is not returnable. View return policy
Description
Description
Proprietary MGB™ and Superbase™ technologies allow post-PCR dissociated melt curve analysis, providing molecular labs with more powerful tools for detecting nucleic acid sequences of specific organisms associated with disease. MGB Alert real-time PCR reagents are optimized for use with most commercially available real-time PCR instruments. With universal cycling conditions, the reagents can easily be incorporated into existing workflow. Laboratories may use MGB Alert reagents to develop real-time PCR assays.
  • Compatible with most real-time PCR instruments
  • Optimized for universal cycling conditions
  • Easy to implement into existing workflow
  • Improved analysis from post-PCR melt curve capabilities
MGB Alert BK Virus Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the VP1 region of BK Virus
MGB Alert CMV Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the IE1 region of CMV
MGB Alert CMV Q Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the IE1 region of CMV
MGB Alert EBV Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the p140 region of the BNRF1 gene of EBV
MGB Alert Enterovirus Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the 5' UTR region of Enterovirus
MGB Alert HHV-6 Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the U67 region of HHV-6
MGB Alert hMPV Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the Polymerase (L) gene of hMPV
MGB Alert HSV Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the glycoprotein D region of HSV
MGB Alert HSV 1 and 2 Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the glycoprotein D region of HSV 1 and 2
MGB Alert MS2 Primers consist of chemically synthesized DNA oligonucleotide primers that amplify nucleic acid sequences from the MS2 bacteriophage
Specifications
Specifications
300μL Chemically synthesized DNA oligonucleotide primers
DNA Sequencing
480
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.