Learn More
ELiTech Molecular Diagnostics Real-Time PCR Internal Controls
Supplier: Nanogen, Inc 800432
Description
Available as FDA-cleared unassayed quality control materials, Real-Time PCR Internal Controls detect variation that may arise during the DNA or RNA extraction and amplification process associated with real-time PCR assays. Each set includes primers and probes that can be used to amplify and detect an internal control template (also included).
All Internal Controls use innovative MGB™ technology, including proprietary probe technology, providing labs with a powerful tool to detect variation in their real-time PCR assays. Real-Time PCR Internal Controls are optimized for use with most commercially available real-time PCR instruments. With universal cycling conditions, the reagents can easily be incorporated into an existing workflow.
- Universal Cycling Conditions: Run multiple assays simultaneously
- Reduced Royalties: Real-time PCR without TaqMan™ probes
- Streamlined Workflow: Bring new molecular tests on quickly
- Open Platform: Runs on most real-time PCR instruments
Specifications
Chemically synthesized DNA oligonucleotide primers and probes | |
CMV 3 Internal Control Set | |
480 | |
Real Time PCR (qPCR) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.