Learn More
ELiTech Molecular Diagnostics Real Time PCR Positive Control Sets
Verify proper performance of an assay and detect problems during the DNA or RNA extraction and amplification process associated with real-time PCR assay
Supplier: Nanogen, Inc M400395
Description
Each Positive Control ASR consists of plasmid containing the target insert associated with the relevant ASR primers and ASR probes product.
- Monitor aspects of PCR testing, lysis, extraction and PCR amplification, and detection
- Estimates testing precision and detects errors in laboratory testing procedures
- Effectively measures assay and instrument integrity
Specifications
One vial of plasmid containing target insert associated with relevant ASR primers and ASR probes | |
BK Virus Positive Control Set | |
24 | |
Real Time PCR (qPCR) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.