Learn More
ELiTech Molecular Diagnostics Real Time PCR Positive Control Sets
Verify proper performance of an assay and detect problems during the DNA or RNA extraction and amplification process associated with real-time PCR assay
Supplier:  Nanogen, Inc M400399
 View more versions of this product
Catalog No. 22-800-239
This item is not returnable. View return policy
Description
Description
Each Positive Control ASR consists of plasmid containing the target insert associated with the relevant ASR primers and ASR probes product.
  • Monitor aspects of PCR testing, lysis, extraction and PCR amplification, and detection
  • Estimates testing precision and detects errors in laboratory testing procedures
  • Effectively measures assay and instrument integrity
Specifications
Specifications
One vial of plasmid containing target insert associated with relevant ASR primers and ASR probes
HSV-1 Positive Control Set
24
Real Time PCR (qPCR)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.