Learn More
Electron Microscopy Sciences Aurion EM Anionic Gold Tracer, EM-grade 25 nm 5 ML
Supplier: Electron Microscopy Sciences 25473
Aurion Gold Tracers are used to visualize charged moieties in specimens: anionic tracers bind to polycationic moieties (basic proteins e.g. histones, cationic tracers bind to polyanionic moieties (membranes, acidic proteins). “Neutral” BSA coated gold tracers are useful for detecting open connections and tissue damage. Under proper conditions and with suitable microscopical techniques the tracers can also be used to follow cellular events in time.
Particle size: EM-grade 25 nm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.