Learn More
Electron Microscopy Sciences Aurion EM Streptavidin/Gold, EM-grade 10nm 2.5 ML
Supplier:  Electron Microscopy Sciences 25268
Encompass_Preferred
Streptavidin conjugated with colloidal gold particles in solution.
Particle size: EM-grade 10nm
Catalog No. 50-291-46
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.