Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Amicon™ Pro Affinity Concentration Kit Protein A with Amicon™ Ultra-0.5 Device
Amicon Pro Affinity Concentration Kit Protein A - A Centrifugal Tool for purifying and concentrating Antibodies.
Specifications
Color | Clear with Pink Cap |
---|---|
Material (Membrane) | Ultracel Low-binding Regenerated Cellulose |
Filtration Area | 1 cm2 |
For Use With (Application) | Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment |
Housing Material | Polypropylene |
Includes:
- 12/pk Amicon Pro
- 12/pk Amicon Ultra
- 3mL Protein A resin (Binding capacity: 20mg human IgG/mL agarose)
- 2 X 66mL Bind/Wash Buffer
- 12mL Elution buffer
- 2.5mL Neutralization buffer
Catalog Number | Mfr. No. | Includes | MWCO | Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | MWCO | Type | Price | Quantity & Availability | |||
ACK5030PA
|
MilliporeSigma™
ACK5030PA |
12/Pk. Amicon Ultra NMWCO 30 kd, 3mL Protein A resin (Binding capacity: 20mg human IgG/mL agarose), 2 x 66mL Bind/Wash Buffer, 12mL Elution buffer, 2.5mL Neutralization buffer | 30 kDa | Amicon Pro Affinity Concentration Kit Protein A with 30 kd Amicon Ultra-0.5 Device | N/A | N/A | N/A | ||
ACK5100PA
|
MilliporeSigma™
ACK5100PA |
12/Pk. Amicon Ultra NMWCO 100 kd, 3mL Protein A resin (Binding capacity: 20mg human IgG/mL agarose), 2 x 66mL Bind/Wash Buffer, 12mL Elution buffer, 2.5mL Neutralization buffer | 100 kDa | Amicon Pro Affinity Concentration Kit Protein A with 100 kd Amicon Ultra-0.5 Device | N/A | N/A | N/A | ||
Description
This kit includes 12/pk Amicon Pro, 12/pk Amicon Ultra NMWCO, 3mL Protein A resin (Binding capacity: 20mg human IgG/ml agarose), 2 X 66ml Bind/Wash Buffer, 12mL Elution buffer, 2.5mL Neutralization bufferOrder Info
Reagents available. (See Cat. No. ACR5000PA)
Specifications
Clear with Pink Cap | |
1 cm2 | |
Polypropylene | |
12/Pk. | |
4.7 in. | |
1.2 in. |
Ultracel Low-binding Regenerated Cellulose | |
Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment | |
4000xg for concentration and buffer exchange spin; 1000xg for flow through, wash, elution and reverse spin | |
10 mL | |
12 cm | |
29.7 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title