Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Chemicon™ alpha/beta Integrin-mediated Cell Adhesion Array Combo Kit, Fluorimetric
For assessing the presence or absence of specific integrins on the cell surface
Supplier: MilliporeSigma™ ECM535
Specifications
alpha/beta-Integrin-mediated Cell Adhesion Array Kit | |
For assessing the presence or absence of specific integrins on the cell surface | |
1/Pk. |
Fluorescence | |
Assay Buffer, alpha/beta Integrin Array Plate, 4X-cell lysis buffer, CyQuant GR Dye |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction