Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ QCM Gelatin Invadopodia Assay, Green

Catalog No. ECM670 Shop All MilliporeSigma Products
Click to view available options
:

Migration, Invasion Assay

Specifications

Content And Storage

Store fluorescein-gelatin, unlabeled gelatin, poly-L-Lysine, TRITC-phalloidin and DAPI at 2-8°C. Store Glutaraldehyde at -20°C. Use all reagents within 4 months.

Description

QCM Gelatin Invadopodia Assay (Green)

Detection Method

Fluorescence

For Use With (Application)

Migration, Invasion Assay

Includes

Glutaraldehyde, 16X, fluorescein-gelatin, 4X, unlabeled gelatin, 4X, poly-L-Lysine, 2X, TRITC-phalloidin, DAPI

Quantity

1 Kit (32 Assays)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.