Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Harleco™ May-Grunwald Solution

Catalog No. 23750044 Shop All MilliporeSigma Products
Click to view available options
:
32 oz.
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

For use with the MIDAS™ III Automated Stainer

Specifications

Quantity 1L
Boiling Point 64.5°C
Format Liquid
Type May-Grunwald Solution
For Use With (Application) Blood stain producing results similar to Wright stain, may be used in conjunction with giemsa stain for bone marrow preparations
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.