Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Harleco™ Phosphate Buffer
Shop All MilliporeSigma Products
Click to view available options
Boiling Point:
100°C
99.9°C
pH:
6.4
6.8
Quantity:
1L
4L
Description
- Used in Wright and Wright-Giemsa staining protocols
Specifications
Specifications
Quantity | 1L |
Boiling Point | 100°C |
Format | Liquid |
Type | Phosphate Buffer |
Color | Colorless |
pH | 6.4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction