Learn More
Azzota Corp Emergency Eye Wash Station With Single Spray Heads, Tuv Certified,Tested According To Us Ansi Z.358.1-2014 Guidelines, H59-1 Brass Chemical Composition Copper Body, 5 Feet Flexible G1/2 Pvc Hose
Supplier: Azzota Corp KC1R