Learn More
Azzota Corp Emergency Eye Wash Station With Two Spray Heads,Tuv Certified,Tested According To Us Ansi Z.358.1-2014 Guidelines,H59-1 Brass Chemical Composition Copper Body, 5 Feet Flexible G1/2 Pvc Hose
Supplier:  Azzota Corp KC2R
Encompass_Preferred
Emergency eye wash station with two spray heads,TUV certified,Tested according to US ANSI Z.358.1-2014 guidelines,H59-1 Brass chemical composition Copper body, 5 feet Flexible G1/2 PVC hose
Catalog No. 50-161-2859
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.