Learn More
AirClean™ Systems Endeavour™ Ductless Fume Hood, Folding Sash
Based on SafeChange technology that minimizes threat of exposure to toxic powders during pre-filter replacement
Supplier: AirClean™ Systems ACPT5000
Alert:
Requires an air filter in order to operate. Air filter must be purchased separately.
Description
Certifications
UL listed, CSA, CE marked
Compliance
Meets all relevant OSHA and ANSI Z9.5 standards
Specifications
Ductless Fume Hood | |
26 x 59 x 35 in. (66 x 149.8 x 89 cm) |