Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Harris' Alum Hematoxylin Stain (Mercury Free with Glacial Acetic Acid)

Catalog No. ES8497
Click to view available options
Quantity:

Harris' Alum Hematoxylin Stain (Mercury Free with Glacial Acetic Acid) is a regressive stain used in demonstrating nuclear chromatin.

  • Staining time: 4 to 8 minutes depending on the nature of tissues
  • Contents: Hematoxylin, Ethanol, Potassium Alum Hematoxylin, Ethanol, Potassium Alum, Glacial Acetic Acid

Specifications

Content And Storage

Hematoxylin, Ethanol, Potassium Alum Hematoxylin, Ethanol, Potassium Alum, Glacial Acetic Acid

Packaging Type

Bottle

Product Type

Hematoxylin

Quantity

1 x 3800 mL (1 gal.)

For Use With (Application)

In vitro diagnostic use only

Test Time

4 to 8 min.

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.