Learn More
NEOGEN Neogen® Environmental Scrub Sampler Stick with Wide Spectrum Neutralizer
SDP
Supplier:  NEOGEN 700002338
Encompass
Neogen® Environmental Scrub Sampler StickSKU No. 700002338 | Catalog No. ESS10WSNMultilayered non-woven composite material maintains integrity when sampling rough areas and scrub dot technology disrupts biofilm and enhances sample collectionStick is metal detectable and features an easy-to-use 1-step twist detachment mechanismNeogen® Wide Spectrum Neutralizer is formulated to effectively neutralize common sanitizers and is designed to be compatible with Neogen® Petrifilm® Plates and Neogen® Molecular Detection System.HACCP International Food Safety Programme Certification I-PE-140-NEO-RG-01
Catalog No. NC3236151
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.